Assay Details
| Gene Symbol: irx4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020493.2 | NP_001018329.2 | 3 - 4 | 824 | 74 | |
| GenBank mRNA | AY845129.1 | - | 3 - 4 | 556 | 74 | |
| BC093262.1 | - | 3 - 4 | 682 | 74 | ||
More Information
Set Membership: |
|

irx4b
Genomic Map