Assay Details
| Gene Symbol: casp6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020497.1 | NP_001018333.1 | 2 - 3 | 155 | 134 | |
| GenBank mRNA | BC094299.1 | - | 2 - 3 | 155 | 134 | |
| BC164245.1 | - | 2 - 3 | 108 | 134 | ||
More Information
Set Membership: |
|

casp6a
Genomic Map