Assay Details
| Gene Symbol: casp6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020497.1 | NP_001018333.1 | 7 - 8 | 744 | 83 | |
| GenBank mRNA | BC094299.1 | - | 7 - 8 | 744 | 83 | |
| BC164245.1 | - | 7 - 8 | 697 | 83 | ||
| MG957996.1 | - | 7 - 8 | 660 | 83 | ||
More Information
Set Membership: |
|

casp6a
Genomic Map