Assay Details
| Gene Symbol: rab3ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020581.2 | NP_001018417.1 | 6 - 7 | 923 | 58 | |
| GenBank mRNA | BC095215.1 | - | 6 - 7 | 924 | 58 | |
More Information
Set Membership: |
|

rab3ip
Genomic Map