Assay Details
| Gene Symbol: ctdsp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009304902.3 | XP_009303177.1 | 5 - 6 | 819 | 68 | |
| XM_021479012.1 | XP_021334687.1 | 4 - 5 | 3407 | 68 | ||
| GenBank mRNA | BC095870.1 | - | 5 - 6 | 758 | 68 | |
More Information
Set Membership: |
|

ctdsp1
Genomic Map