Assay Details
| Gene Symbol: sla1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034989.2 | NP_001030161.1 | 7 - 8 | 793 | 73 | |
| GenBank mRNA | BC103485.1 | - | 7 - 8 | 797 | 73 | |
More Information
Set Membership: |
|

sla1a
Genomic Map