Assay Details
| Gene Symbol: vegfd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040178.1 | NP_001035268.1 | 2 - 3 | 285 | 63 | |
| XM_005169310.4 | XP_005169367.1 | 2 - 3 | 986 | 63 | ||
| XM_009306087.3 | XP_009304362.1 | 2 - 3 | 1110 | 63 | ||
| GenBank mRNA | BC163164.1 | - | 2 - 3 | 338 | 63 | |
| BC163170.1 | - | 2 - 3 | 338 | 63 | ||
| DQ402074.1 | - | 2 - 3 | 285 | 63 | ||
More Information
Set Membership: |
|

vegfd
Genomic Map