Assay Details
| Gene Symbol: sult4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040244.1 | NP_001035334.1 | 3 - 4 | 471 | 69 | |
| GenBank mRNA | BC115282.1 | - | 3 - 4 | 471 | 69 | |
More Information
Set Membership: |
|

sult4a1
Genomic Map