Assay Details
| Gene Symbol: jazf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044955.1 | NP_001038420.1 | 4 - 5 | 545 | 105 | |
| XM_005159518.4 | XP_005159575.1 | 4 - 5 | 3268 | 105 | ||
More Information
Set Membership: |
|

jazf1a
Genomic Map