Assay Details
| Gene Symbol: batf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045392.1 | NP_001038857.1 | 2 - 3 | 179 | 60 | |
| GenBank mRNA | BC122303.1 | - | 2 - 3 | 179 | 60 | |
| BC164923.1 | - | 2 - 3 | 210 | 60 | ||
More Information
Set Membership: |
|

batf3
Genomic Map