Assay Details
| Gene Symbol: ccdc126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045403.1 | NP_001038868.1 | 2 - 3 | 692 | 67 | |
| GenBank mRNA | BC122382.1 | - | 2 - 3 | 692 | 67 | |
More Information
Set Membership: |
|

ccdc126
Genomic Map