Assay Details
| Gene Symbol: neurl1aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077558.2 | NP_001071026.1 | 3 - 4 | 733 | 81 | |
| XM_021475292.1 | XP_021330967.1 | 3 - 4 | 850 | 81 | ||
| GenBank mRNA | BC125926.1 | - | 3 - 4 | 733 | 81 | |
More Information
Set Membership: |
|

neurl1aa
Genomic Map