Assay Details
| Gene Symbol: nsd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082551.1 | NP_001076020.1 | 10 - 11 | 2342 | 84 | |
| GenBank mRNA | DQ358102.1 | - | 9 - 10 | 1805 | 84 | |
More Information
Set Membership: |
|

nsd2
Genomic Map