Assay Details
| Gene Symbol: nsd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082551.1 | NP_001076020.1 | 18 - 19 | 3514 | 113 | |
| GenBank mRNA | BC115211.1 | - | 3 - 4 | 335 | 113 | |
| BC171696.1 | - | 18 - 19 | 3508 | 113 | ||
| DQ358102.1 | - | 17 - 18 | 2977 | 113 | ||
More Information
Set Membership: |
|

nsd2
Genomic Map