Assay Details
| Gene Symbol: nsd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082551.1 | NP_001076020.1 | 6 - 7 | 1902 | 64 | |
| GenBank mRNA | BC171696.1 | - | 6 - 7 | 1896 | 64 | |
More Information
Set Membership: |
|

nsd2
Genomic Map