Assay Details
| Gene Symbol: jag2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131862.1 | NP_571937.1 | 18 - 19 | 2516 | 103 | |
| XM_005160485.4 | XP_005160542.1 | 17 - 18 | 2423 | 103 | ||
| GenBank mRNA | AF090432.1 | - | 18 - 19 | 2499 | 103 | |
| AF229449.1 | - | 18 - 19 | 2516 | 103 | ||
| AF229450.1 | - | 17 - 18 | 2402 | 103 | ||
| BC163889.1 | - | 18 - 19 | 2503 | 103 | ||
| CU458993.1 | - | 18 - 19 | 2499 | 103 | ||
More Information
Set Membership: |
|

jag2b
Genomic Map