Assay Details
| Gene Symbol: jag2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131862.1 | NP_571937.1 | 19 - 20 | 2541 | 66 | |
| XM_005160485.4 | XP_005160542.1 | 18 - 19 | 2448 | 66 | ||
| GenBank mRNA | AF090432.1 | - | 19 - 20 | 2524 | 66 | |
| AF229449.1 | - | 19 - 20 | 2541 | 66 | ||
| AF229450.1 | - | 18 - 19 | 2427 | 66 | ||
| BC163889.1 | - | 19 - 20 | 2528 | 66 | ||
| CU458993.1 | - | 19 - 20 | 2524 | 66 | ||
More Information
Set Membership: |
|

jag2b
Genomic Map