Assay Details
| Gene Symbol: jag2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131862.1 | NP_571937.1 | 21 - 22 | 2750 | 60 | |
| XM_005160485.4 | XP_005160542.1 | 20 - 21 | 2657 | 60 | ||
| GenBank mRNA | AF090432.1 | - | 21 - 22 | 2733 | 60 | |
| AF229449.1 | - | 21 - 22 | 2750 | 60 | ||
| AF229450.1 | - | 20 - 21 | 2636 | 60 | ||
| BC163889.1 | - | 21 - 22 | 2737 | 60 | ||
| CU458993.1 | - | 21 - 22 | 2733 | 60 | ||
More Information
Set Membership: |
|

jag2b
Genomic Map