Assay Details
| Gene Symbol: jag2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131862.1 | NP_571937.1 | 23 - 24 | 3111 | 73 | |
| XM_005160485.4 | XP_005160542.1 | 22 - 23 | 3018 | 73 | ||
| GenBank mRNA | AF090432.1 | - | 23 - 24 | 3094 | 73 | |
| AF229449.1 | - | 23 - 24 | 3111 | 73 | ||
| AF229450.1 | - | 22 - 23 | 2997 | 73 | ||
| BC163889.1 | - | 23 - 24 | 3098 | 73 | ||
| CU458993.1 | - | 23 - 24 | 3094 | 73 | ||
More Information
Set Membership: |
|

jag2b
Genomic Map