Assay Details
| Gene Symbol: alas2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131682.2 | NP_571757.1 | 5 - 6 | 664 | 55 | |
| GenBank mRNA | AF095747.1 | - | 5 - 6 | 726 | 55 | |
| BC056338.1 | - | 5 - 6 | 664 | 55 | ||
| BC067149.1 | - | 5 - 6 | 683 | 55 | ||
| CF943699.1 | - | 3 - 4 | 434 | 55 | ||
More Information
Set Membership: |
|

alas2
Genomic Map