Assay Details
| Gene Symbol: pdia4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199779.2 | NP_956073.2 | 4 - 5 | 849 | 114 | |
| GenBank mRNA | BC045862.1 | - | 4 - 5 | 114 | ||
| BC063979.1 | - | 4 - 5 | 114 | |||
| BC117629.1 | - | 4 - 5 | 752 | 114 | ||
| BC165633.1 | - | 4 - 5 | 114 | |||
More Information
Set Membership: |
|

pdia4
Genomic Map