Assay Details
| Gene Symbol: ranbp3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200690.1 | NP_956984.1 | 3 - 4 | 142 | 82 | |
| XM_005168445.4 | XP_005168502.1 | 3 - 4 | 169 | 82 | ||
| GenBank mRNA | BC059200.1 | - | 3 - 4 | 142 | 82 | |
More Information
Set Membership: |
|

ranbp3b
Genomic Map