Assay Details
| Gene Symbol: gnao1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200787.2 | NP_957081.1 | 4 - 5 | 815 | 62 | |
| XM_005159069.4 | XP_005159126.1 | 4 - 5 | 1055 | 62 | ||
| XM_005159071.4 | XP_005159128.1 | 4 - 5 | 1053 | 62 | ||
| GenBank mRNA | BC059637.1 | - | 4 - 5 | 751 | 62 | |
More Information
Set Membership: |
|

gnao1a
Genomic Map