Assay Details
| Gene Symbol: gnao1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200787.2 | NP_957081.1 | 7 - 8 | 1238 | 58 | |
| GenBank mRNA | BC059637.1 | - | 7 - 8 | 1174 | 58 | |
More Information
Set Membership: |
|

gnao1a
Genomic Map