Assay Details
| Gene Symbol: ccdc58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200807.1 | NP_957101.1 | 2 - 3 | 179 | 102 | |
| XM_005162679.4 | XP_005162736.1 | 2 - 3 | 221 | 102 | ||
| GenBank mRNA | BC059679.1 | - | 2 - 3 | 179 | 102 | |
More Information
Set Membership: |
|

ccdc58
Genomic Map