Assay Details
| Gene Symbol: rnf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205553.1 | NP_991116.1 | 1 - 2 | 179 | 81 | |
| NM_205766.1 | NP_991329.1 | 1 - 2 | 232 | 81 | ||
| GenBank mRNA | BC044545.1 | - | 1 - 2 | 232 | 81 | |
| BC065643.1 | - | 1 - 2 | 179 | 81 | ||
More Information
Set Membership: |
|

rnf8
Genomic Map