Assay Details
| Gene Symbol: otub1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002500.2 | NP_001002500.2 | 3 - 4 | 227 | 89 | |
| GenBank mRNA | BC076301.1 | - | 3 - 4 | 215 | 89 | |
| EH433266.1 | - | 3 - 4 | 253 | 89 | ||
More Information
Set Membership: |
|

otub1b
Genomic Map