Assay Details
| Gene Symbol: mrps23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003615.1 | NP_001003615.1 | 1 - 2 | 158 | 63 | |
| XM_005157688.4 | XP_005157745.1 | 1 - 2 | 185 | 63 | ||
| GenBank mRNA | BC077090.1 | - | 1 - 2 | 158 | 63 | |
More Information
Set Membership: |
|

mrps23
Genomic Map