Assay Details
| Gene Symbol: colec11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007331.1 | NP_001007332.1 | 3 - 4 | 353 | 82 | |
| XM_009293191.3 | XP_009291466.1 | 3 - 4 | 82 | |||
| XM_009293192.3 | XP_009291467.1 | 3 - 4 | 82 | |||
| GenBank mRNA | BC085557.1 | - | 3 - 4 | 353 | 82 | |
More Information
Set Membership: |
|

colec11
Genomic Map