Assay Details
| Gene Symbol: sept10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017557.1 | NP_001017557.1 | 6 - 7 | 869 | 70 | |
| XM_021478648.1 | XP_021334323.1 | 6 - 7 | 911 | 70 | ||
| GenBank mRNA | BC092936.1 | - | 6 - 7 | 869 | 70 | |
More Information
Set Membership: |
|

sept10
Genomic Map