Assay Details
| Gene Symbol: lims2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020670.1 | NP_001018506.1 | 1 - 2 | 149 | 81 | |
| XM_021478667.1 | XP_021334342.1 | 1 - 2 | 184 | 81 | ||
| GenBank mRNA | BC065908.1 | - | 1 - 2 | 135 | 81 | |
| BC095708.1 | - | 1 - 2 | 149 | 81 | ||
More Information
Set Membership: |
|

lims2
Genomic Map