Assay Details
| Gene Symbol: bcl2l13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044891.2 | NP_001038356.1 | 6 - 7 | 826 | 74 | |
| XM_005159108.4 | XP_005159165.1 | 6 - 7 | 798 | 74 | ||
| XM_017351933.2 | XP_017207422.1 | 5 - 6 | 643 | 74 | ||
| XM_021467663.1 | XP_021323338.1 | 6 - 7 | 807 | 74 | ||
| GenBank mRNA | BC139852.1 | - | 6 - 7 | 826 | 74 | |
More Information
Set Membership: |
|

bcl2l13
Genomic Map