Assay Details
| Gene Symbol: trim62.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080674.1 | NP_001074143.1 | 2 - 3 | 680 | 82 | |
| XM_021479666.1 | XP_021335341.1 | 2 - 3 | 712 | 82 | ||
| GenBank mRNA | BC129354.1 | - | 2 - 3 | 680 | 82 | |
More Information
Set Membership: |
|

trim62.1
Genomic Map