Assay Details
| Gene Symbol: trim62.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080674.1 | NP_001074143.1 | 5 - 6 | 1150 | 104 | |
| XM_021479666.1 | XP_021335341.1 | 6 - 7 | 1227 | 104 | ||
| GenBank mRNA | BC129354.1 | - | 5 - 6 | 1150 | 104 | |
| Gene Symbol: trim62.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021479901.1 | XP_021335576.1 | 4 - 5 | 510 | 104 | |
More Information
Set Membership: |
|

trim62.1
Genomic Map