Assay Details
| Gene Symbol: prkra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089361.1 | NP_001082830.1 | 4 - 5 | 402 | 66 | |
| XM_005172508.4 | XP_005172565.1 | 4 - 5 | 386 | 66 | ||
| GenBank mRNA | BC135034.1 | - | 4 - 5 | 402 | 66 | |
More Information
Set Membership: |
|

prkra
Genomic Map