Assay Details
| Gene Symbol: pcgf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089369.1 | NP_001082838.1 | 6 - 7 | 736 | 88 | |
| GenBank mRNA | BC139554.1 | - | 6 - 7 | 736 | 88 | |
More Information
Set Membership: |
|

pcgf6
Genomic Map