Assay Details
| Gene Symbol: ugdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110402.1 | NP_001103872.1 | 4 - 5 | 562 | 85 | |
| GenBank mRNA | AF361478.1 | - | 3 - 4 | 458 | 85 | |
| BC154794.1 | - | 4 - 5 | 562 | 85 | ||
| BC163406.1 | - | 3 - 4 | 546 | 85 | ||
| BC163432.1 | - | 3 - 4 | 546 | 85 | ||
More Information
Set Membership: |
|

ugdh
Genomic Map