Assay Details
| Gene Symbol: jak2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131093.1 | NP_571168.1 | 17 - 18 | 2364 | 57 | |
| XM_009295057.3 | XP_009293332.1 | 17 - 18 | 2542 | 57 | ||
| GenBank mRNA | AB018222.1 | - | 14 - 15 | 1745 | 57 | |
| AJ005690.1 | - | 17 - 18 | 2364 | 57 | ||
| BC162596.1 | - | 17 - 18 | 2323 | 57 | ||
More Information
Set Membership: |
|

jak2a
Genomic Map