Assay Details
| Gene Symbol: jak2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131093.1 | NP_571168.1 | 11 - 12 | 1605 | 100 | |
| XM_009295057.3 | XP_009293332.1 | 11 - 12 | 1783 | 100 | ||
| GenBank mRNA | AJ005690.1 | - | 11 - 12 | 1605 | 100 | |
| BC162596.1 | - | 11 - 12 | 1564 | 100 | ||
More Information
Set Membership: |
|

jak2a
Genomic Map