Assay Details
| Gene Symbol: cyp26a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131146.2 | NP_571221.2 | 6 - 7 | 1213 | 88 | |
| GenBank mRNA | BC055232.1 | - | 6 - 7 | 1213 | 88 | |
More Information
Set Membership: |
|

cyp26a1
Genomic Map