Assay Details
| Gene Symbol: tbxta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131162.1 | NP_571237.1 | 4 - 5 | 732 | 60 | |
| GenBank mRNA | BC162297.1 | - | 4 - 5 | 727 | 60 | |
| BC162300.1 | - | 4 - 5 | 727 | 60 | ||
| S57147.1 | - | 4 - 5 | 732 | 60 | ||
More Information
Set Membership: |
|

tbxta
Genomic Map