Assay Details
| Gene Symbol: bmp7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131321.2 | NP_571396.1 | 1 - 2 | 583 | 81 | |
| GenBank mRNA | AF201379.1 | - | 1 - 2 | 584 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

bmp7a| Gene Symbol: bmp7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131321.2 | NP_571396.1 | 1 - 2 | 583 | 81 | |
| GenBank mRNA | AF201379.1 | - | 1 - 2 | 584 | 81 | |
Set Membership: |
|