Assay Details
| Gene Symbol: ccdc43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200658.2 | NP_956952.1 | 1 - 2 | 208 | 68 | |
| GenBank mRNA | BC057502.1 | - | 1 - 2 | 210 | 68 | |
| BC155274.1 | - | 1 - 2 | 256 | 68 | ||
More Information
Set Membership: |
|

ccdc43
Genomic Map