Assay Details
| Gene Symbol: uqcrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200820.1 | NP_957114.1 | 1 - 2 | 54 | 66 | |
| GenBank mRNA | BC059705.1 | - | 1 - 2 | 54 | 66 | |
| BC070011.1 | - | 1 - 2 | 59 | 66 | ||
More Information
Set Membership: |
|

uqcrc1
Genomic Map