Assay Details
| Gene Symbol: prkg1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201030.1 | NP_957324.1 | 4 - 5 | 895 | 71 | |
| XM_005157058.4 | XP_005157115.1 | 3 - 4 | 929 | 71 | ||
| GenBank mRNA | BC054581.1 | - | 4 - 5 | 895 | 71 | |
More Information
Set Membership: |
|

prkg1a
Genomic Map