Assay Details
| Gene Symbol: cbr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205656.2 | NP_991219.2 | 4 - 5 | 505 | 97 | |
| GenBank mRNA | BC065615.1 | - | 4 - 5 | 486 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cbr4| Gene Symbol: cbr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205656.2 | NP_991219.2 | 4 - 5 | 505 | 97 | |
| GenBank mRNA | BC065615.1 | - | 4 - 5 | 486 | 97 | |
Set Membership: |
|