Assay Details
| Gene Symbol: dao.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205694.1 | NP_991257.1 | 3 - 4 | 355 | 63 | |
| XM_021476050.1 | XP_021331725.1 | 2 - 3 | 314 | 63 | ||
| GenBank mRNA | BC065882.1 | - | 3 - 4 | 355 | 63 | |
More Information
Set Membership: |
|

dao.3
Genomic Map