Assay Details
| Gene Symbol: irf2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001326712.1 | NP_001313641.1 | 2 - 3 | 328 | 115 | |
| NM_207054.2 | NP_996937.2 | 2 - 3 | 328 | 115 | ||
| GenBank mRNA | BC066487.1 | - | 2 - 3 | 225 | 115 | |
More Information
Set Membership: |
|

irf2a
Genomic Map