Assay Details
| Gene Symbol: ak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212596.1 | NP_997761.1 | 3 - 4 | 394 | 62 | |
| XM_005159648.3 | XP_005159705.1 | 3 - 4 | 428 | 62 | ||
| GenBank mRNA | BC053160.1 | - | 3 - 4 | 394 | 62 | |
| KJ651951.1 | - | 3 - 4 | 336 | 62 | ||
More Information
Set Membership: |
|

ak2
Genomic Map