Assay Details
| Gene Symbol: rbmx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212598.1 | NP_997763.1 | 3 - 4 | 340 | 65 | |
| XM_017358984.2 | XP_017214473.1 | 3 - 4 | 364 | 65 | ||
| XM_021480965.1 | XP_021336640.1 | 3 - 4 | 364 | 65 | ||
| XM_021480966.1 | XP_021336641.1 | 3 - 4 | 364 | 65 | ||
| GenBank mRNA | AJ717349.2 | - | 2 - 3 | 233 | 65 | |
| AY394938.1 | - | 3 - 4 | 330 | 65 | ||
| BC049509.1 | - | 3 - 4 | 340 | 65 | ||
| BC071326.1 | - | 2 - 3 | 256 | 65 | ||
More Information
Set Membership: |
|

rbmx
Genomic Map